Merck
Z742986
Julabo® CORIO C open heating bath circulator
C-B17, capacity 13 L (17 L), stainless steel bath (Insulated), temperature 20-100 °C, AC/DC input 230 V AC, Schuko plug
Größe auswählen
1 EA
1.470,00 €
1.470,00 €
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
1.470,00 €
About This Item
UNSPSC-Code:
41103706
NACRES:
NB.77
1.470,00 €
Check Cart for Availability
Materialien
stainless steel bath (Insulated)
Verpackung
pack of 1 ea
Hersteller/Markenname
Julabo 9011417
Temperaturmessbereich
20-100 °C
Kapazität
13 L (17 L)
Außenmaß B × L × H
38 cm × 40 cm × 47 cm
Öffnung B × L × H
30 cm × 18 cm × 20 cm
Allgemeine Beschreibung
Open heating bath circulators are designed for internal applications in the circulator bath.
CORIO C open heating bath circulators in this range are equipped with bath tanks made of Polycarbonat or stainless steel.
Available accessories include a variety of test tube racks, immersion-height adjustable platforms and cooling coils. The circulator can be removed for easy cleaning of the bath tanks.
Rechtliche Hinweise
CORIO is a trademark of Julabo GmbH
Julabo is a registered trademark of Julabo USA, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)