Merck
Z648183
Grid coating pen
provides a thin layer of adhesive
Größe auswählen
1 EA
66,40 €
66,40 €
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
66,40 €
About This Item
UNSPSC-Code:
41100000
NACRES:
MA.01
66,40 €
Check Cart for Availability
Hersteller/Markenname
1556
Allgemeine Beschreibung
A light touch with this pen will provide a thin layer of adhesive on the grid. The layer will dry in 1-2 minutes at room temperature and is then ready to receive the sections. Pen may also be used as pre-treatment before coating grids with formvar or other thin films.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)