Merck
Z619094
Heidolph Rotationsverdampfer, Laborota 4003
vertical condenser G3, glass
About This Item
UNSPSC-Code:
41100000
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Aufnahme AC/DC
230 V
Materialien
glass
Leistungsmerkmale
CE compliant
Hersteller/Markenname
Heidolph
Parameter
20-180 °C temperature
Allgemeine Beschreibung
Safety features
  • For your protection against splashing water and steam, the operating panel is located in front of the evaporator for safe and easy access.
  • Drip shield over display protects user and control panel from spills
  • Individual power switches for control panel and heating bath offers additional safety
  • Heating bath features non-slip safety handles for safe and easy transport of bath
  • Heating bath is constructed of an insulated double wall for you protection against burns and scalding
  • Adjustable sliding safety guard hood always aligns with the bath and protects you in case of implosion
Convenience features
  • Long life graphite filled PTFE vacuum seal
  • Non-sticking vapor tube removes easily eliminating broken glass
  • Unique large bore vapor tube offers fast distribution rates
  • One piece vapor tube without additional sealing
  • Flask ejector presses off sticking evaporation flasks
  • Adjust and fix the height of your evaporating flask easily
  • For your convenience, the angle of the evaporation flask can be adjusted from 10 ° to 85 ° quickly
  • Superior results due to high torque (140 Ncm) drive
  • Choice of manual or motor lift
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
Artikel
A complete reversed phase HPLC-ELSD workflow, including sample preparation for the analysis of Polysorbate 80 in a monoclonal antibody formulation.