Merck
Z544701
Molymod® Individual organic model set
Starch or cellulose
Größe auswählen
1 SET
25,20 €
25,20 €
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 SET
25,20 €
About This Item
UNSPSC-Code:
60104506
NACRES:
NB.17
25,20 €
Check Cart for Availability
Beschreibung
glucose units 3
Hersteller/Markenname
Molymod
Allgemeine Beschreibung
The mushroom-shaped hydrogen Atomlink components and the short (non-visible) bonding links provide compact, semi-space-filling models with an internuclear scale of 1.45 cm per Angstrom. Polymer models include long, flexible links for demonstration of the mechanism of polymerization.
Rechtliche Hinweise
Atomlink is a trademark of Spiring Enterprises Ltd.
Molymod is a registered trademark of Spiring Enterprises Ltd.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)