Merck
Z203246
Sigma-Aldrich
Aldrich® anticlimb splash-guard adapter
female joint: ST/NS 29/32, male joint: ST/NS 29/32, capacity 250 mL
Größe auswählen
1 EA
137,90 €
137,90 €
Listenpreis197,00 €Sparen Sie 30%
Versandbereit am22. April 2025Details
Größe auswählen
Ansicht ändern
1 EA
137,90 €
About This Item
UNSPSC-Code:
41121815
NACRES:
NB.35
137,90 €
Listenpreis197,00 €Sparen Sie 30%
Versandbereit am22. April 2025Details
Verbindung
female joint: ST/NS29/32
male joint: ST/NS29/32
Kapazität
250 mL
Verwandte Kategorien
Allgemeine Beschreibung
Aldrich® anticlimb splash-guard adapter greatly lower carry-over from foaming or bumping. Because the venting hole is located at the top, the foam can be prevented from climbing higher into the condenser.
Anwendung
Aldrich® anticlimb splash-guard adapter are ideal for use with solvents that foam excessively during rotary evaporation. Inner vapor tube is sealed to the top of trap to prevent foam entry into condenser.
Rechtliche Hinweise
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)