Merck
Z115738
Sigma-Aldrich
Aldrich® rotary evaporator cleaning adapter
joint: ST/NS 24/40
Größe auswählen
1 EA
80,80 €
80,80 €
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
80,80 €
About This Item
UNSPSC-Code:
41121815
NACRES:
NB.35
80,80 €
Check Cart for Availability
Verbindung
joint: ST/NS24/40
Verwandte Kategorien
Allgemeine Beschreibung
For fast and easy cleaning of evaporator condenser assembly.
Evaporator Cleaning Technique
1. Attach the cleaning adapter to a rotary evaporator.
2. Plug the end of the cleaning adapter.
3. Pull solvent into the system through the feed tube.
4. Quickly release the vacuum.
5. Solvent is sprayed throughout the entire assembly cleaning it quickly and efficiently.
Rechtliche Hinweise
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)