Merck
TEM-CF200F1CU
Sigma-Aldrich
Carbon Film Supported Copper Gilder Finder F1 Grids
size 200 mesh, box of 25
Synonym(e):
CARBON FILM ONLY ON GILDER FI FINDER 200# CU 25/BX
About This Item
UNSPSC-Code:
41111761
NACRES:
MA.24
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Verpackung
box of 25
Größe
200 mesh
Allgemeine Beschreibung
A 5-6 nm thin film of pure carbon supported glider finder (F1) grids. Copper grid, 200 mesh. Gilder finder grids are locator grids with reference/index on the grid: 3.05mm diameter, 0.7 mil (18μm) thickness. F1 type of design has thick bars dividing regions into 6 thin bar areas, which are identified by a numeric system.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)