Merck
930369
Sigma-Aldrich
TEM Grids
Carbon Film Supported Copper Grid, Standard Thickness, grid size 200 mesh, box of 50
Synonym(e):
TEM grids
About This Item
UNSPSC-Code:
41111761
NACRES:
MA.24
product name
Carbon Film Supported Copper Grid, Standard Thickness, grid size 200 mesh, box of 50
Materialien
carbon film
copper grid
Verpackung
box of 50
film size
5-6 nm
Größe des Netzaufdrucks
200 mesh
Allgemeine Beschreibung
A 5-6 nm thin film of pure carbon supported standard square mesh grids.
TEM grids supported by carbon film only. A 5-6 nm thin film of pure carbon deposited on one side of the standard square mesh grid.