Merck
SYNM300005
Synthware all-glass vacuum/inert gas manifold with solid high vacuum stopcocks
number of ports, 5, Hose Connections: Front-right, Rear-left-right
Größe auswählen
1 EA
913,00 €
913,00 €
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
913,00 €
About This Item
UNSPSC-Code:
40161501
NACRES:
NB.51
913,00 €
Check Cart for Availability
Leistungsmerkmale
number of ports 5
Verpackung
pack of 1 ea
Hersteller/Markenname
M300005
Allgemeine Beschreibung
Synthware glass manifold featuring 5 ports, high vacuum solid glass stopcocks that are precision ground with corresponding numbers on the barrel and plug. The solid glass stopcocks have constant pressure retention devices. The beams have a 22 mm O.D. with front-right and rear-left 10 mm hose connections. The distance between the stopcocks is 75 mm center to center.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)