Merck
SYNM270005
Synthware double manifold with hollow high vacuum stopcocks and SJ 35/20 joint
number of ports, 5, joint: ST/NS 35/20, Connections: front-left-right, rear-right
Synonym(e):
Synthware M270005, vacuum manifold, vacuum unit
Größe auswählen
1 EA
728,00 €
728,00 €
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
728,00 €
About This Item
UNSPSC-Code:
40161501
NACRES:
NB.51
728,00 €
Check Cart for Availability
Verbindung
joint: ST/NS35/20
Leistungsmerkmale
number of ports 5
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware M270005
Allgemeine Beschreibung
With 35/20 ball joints and 4mm double oblique high vacuum hollow glass stopcocks precision ground with corresponding numbers on barrel and plugs. Lower vacuum chambers on stopcock are evacuated with a separate hole in the plug in order to hold the plug in the stopcock barrels.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)