Merck
SYNH101410
Synthware Hickman still head with cap, o-ring and PTFE-faced septum
joint: ST/NS 14/10
Synonym(e):
Synthware H101410, Hickman Distillation head
Größe auswählen
1 EA
47,80 €
47,80 €
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
47,80 €
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.51
47,80 €
Check Cart for Availability
Verbindung
joint: ST/NS14/10
Hersteller/Markenname
H101410
Verwandte Kategorien
Allgemeine Beschreibung
Synthware Hickman distillation head is designed for microscale fractional distillation. It is supplied with one compression cap, one Viton® O-ring and one PTFE - faced silicone rubber septum. Replacement accessories available.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)