Merck
MT11124303/EU
Mettler Toledo Printer RS-P26, RS232 interface
AC/DC input 230 V AC, EuroPlug
Größe auswählen
1 EA
1.040,00 €
1.040,00 €
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
1.040,00 €
About This Item
UNSPSC-Code:
41115600
NACRES:
NB.61
1.040,00 €
Check Cart for Availability
Hersteller/Markenname
Mettler-Toledo 11124303/EU
Allgemeine Beschreibung
The RS-P26 Compact Printer generates GxP-compliant printouts on regular or adhesive paper. The ergonomic design makes operation easy and allows tarring and printing of the balance directly from the printer. Thanks to the compact footprint, the RS-P26 printer finds place even on smallest work places.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)