Merck
CLS56001L
Pyrex® Lifetime-Red volumetric flask with snap cap
capacity 1,000 mL
Synonym(e):
clear flasks, pyrex flask, volumetric flasks, Corning® 5600
About This Item
UNSPSC-Code:
41121804
NACRES:
NB.43
Materialien
borosilicate glass
long
Sterilität
non-sterile
Leistungsmerkmale
closure type Snap cap
Verpackung
case of 6
Hersteller/Markenname
Corning 5600-1L
tol. measuring range
0.30 mL
Kapazität
1,000 mL
Allgemeine Beschreibung
Capacity mark is fine line-etched through narrow, red band into clear glass.
Rechtliche Hinweise
Corning is a registered trademark of Corning, Inc.
Lifetime-Red is a trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)