Merck
CLS44464L
Pyrex® shaker flask with extra-deep baffles
capacity 4,000 mL
Synonym(e):
Corning® 4446
About This Item
UNSPSC-Code:
41121804
NACRES:
NB.43
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Materialien
Delong
borosilicate glass
Sterilität
non-sterile
Leistungsmerkmale
cap: no
closure type Closure cap
Verpackung
case of 2
Hersteller/Markenname
Corning 4446-4L
Kapazität
4,000 mL
Allgemeine Beschreibung
Delong style neck, with three extra-deep baffles, to enhance gas transfer when used with rotary or reciprocating shakers. Long neck reduces splashing and is designed for two-position polypropylene closures. Closures are sold separately.
Rechtliche Hinweise
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)