Merck
BR781565
BRAND® full rack with lid and tubes
Includes 96 coded 0.65 mL tubes
Synonym(e):
Sample Storage, Tube racks, Tubes
Größe auswählen
50 EA
656,00 €
656,00 €
Check Cart for Availability
Größe auswählen
Ansicht ändern
50 EA
656,00 €
About This Item
UNSPSC-Code:
41122804
NACRES:
NB.12
656,00 €
Check Cart for Availability
Materialien
polypropylene
Leistungsmerkmale
Holds 96 x 0.65 mL tubes
Verpackung
pack of 50 ea
Hersteller/Markenname
BRAND 781565
Allgemeine Beschreibung
Tubes and rack are autoclavable at 121 °C (2 bar), acc. DIN EN 285. Cover mat / strips (TPE) not autoclavable. Tubes, racks, and lids can withstand temperatures as low as –80 °C. The seal of the tubes is pressure-tested. Thanks to the stable connection between the mounting plate and the racks, the stackable SBS racks are especially well-suited for use with robots and other automated dispensing systems. Coded tubes (A1 – H12) and rack sides suitable for barcode labels simplify sample organization.
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)