Merck
BR759235
BRAND® UV cuvette micro
center H 15 mm, volume 70-550 μL, pack of 100 ea (individually wrapped)
Synonym(e):
Cuvettes, UV-Cuvettes
Größe auswählen
100 EA
61,80 €
61,80 €
Versandbereit am28. April 2025Details
Größe auswählen
Ansicht ändern
100 EA
61,80 €
About This Item
UNSPSC-Code:
41121813
NACRES:
SB.32
61,80 €
Versandbereit am28. April 2025Details
Verpackung
pack of 100 ea (individually wrapped)
Hersteller/Markenname
BRAND 759235
Mitte H
15 mm
Volumen
70-550 μL
Allgemeine Beschreibung
  • BRAND® UV cuvette micro is manufactured under controlled room conditions.
  • Ideal for the determinations in water analysis, chemistry, and life science applications.
  • Cuvette material offers high chemical resistance to most polar solvents, acids, and alkaline solutions.
  • Clear, clean optical path with an indication of optical path orientation.
  • Eliminates the risk of cross-contamination.
  • Grouped by mold cavity number.
  • An economical alternative to Quartz cuvettes.
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
Kunden haben sich ebenfalls angesehen
Slide 1 of 1