BR759170
BRAND® UV cuvette
macro, volume 2.5-4.5 mL
Synonym(e):
Cuvettes, UV-Cuvettes
Größe auswählen
36,00 €
Auf LagerDetails
Größe auswählen
About This Item
36,00 €
Auf LagerDetails
Empfohlene Produkte
Hersteller/Markenname
BRAND 759170
Volumen
2.5-4.5 mL
Allgemeine Beschreibung
- BRAND® UV cuvette is manufactured under controlled room conditions.
- Ideal for the determinations in water analysis, chemistry, and life science applications.
- Cuvette material offers high chemical resistance to most polar solvents, acids, and alkaline solutions.
- Clear, clean optical path with an indication of optical path orientation.
- Eliminates the risk of cross-contamination.
- Grouped by mold cavity number.
- An economical alternative to Quartz cuvettes.
Rechtliche Hinweise
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)