Merck
BR722060
BRAND® sample vial
for Leuko-Ery Counter, capacity 12 mL, with lid
Synonym(e):
Sample Cups
Größe auswählen
1000 EA
166,00 €
166,00 €
Versandbereit am22. April 2025Details
Größe auswählen
Ansicht ändern
1000 EA
166,00 €
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.11
166,00 €
Versandbereit am22. April 2025Details
Materialien
colorless tube
flat bottom
flat cap
polyethylene cap
polystyrene
polystyrene tube
white cap
Sterilität
non-sterile
Leistungsmerkmale
graduations: no
Verpackung
pack of 1000 ea
Hersteller/Markenname
BRAND 722060
Kapazität
12 mL
Allgemeine Beschreibung
The BRAND® Sample Vial is designed for secure and efficient storage of samples in laboratory settings. Made from high-quality transparent polystyrene (PS), this vial provides excellent visibility for easy identification and monitoring of contents. It features a snap cap made from polyethylene (PE), ensuring a tight seal that protects samples from contamination and evaporation.
Leistungsmerkmale und Vorteile
  • BRAND® sample vial is designed for various laboratory applications.
  • Made of transparent polystyrene for accurate and quick sample analysis.
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)