Merck
BR143270
BRAND® special laboratory tubing
I.D. × O.D. × W 6 mm × 9 mm × 1.5 mm, L 20 m
Synonym(e):
Special laboratory tubing, Tubing
About This Item
UNSPSC-Code:
41105108
NACRES:
NB.35
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Materialien
PVC
Hersteller/Markenname
BRAND 143270
ID × AD × B
6 mm × 9 mm × 1.5 mm
Länge
20 m
Allgemeine Beschreibung
Particularly high-grade material, manufactured to a first-class standard. Good flexibility and chemical resistance make this tubing ideal for use with apparatus assemblies. Autoclavable (121 °C).
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)