Merck
BR113125
BRAND® forcep with rectangular ends
PTFE, L 100 mm
Synonym(e):
Forceps, PTFE
About This Item
UNSPSC-Code:
41122405
NACRES:
NB.35
Materialien
PTFE
Hersteller/Markenname
BRAND 113125
Länge
100 mm
Allgemeine Beschreibung
The BRAND® Forceps with Rectangular Ends are specialized tools designed for handling materials in challenging laboratory environments. Made from high-quality PTFE (polytetrafluoroethylene), these forceps offer exceptional durability and performance. The PTFE construction ensures excellent resistance to a wide range of chemicals, making these forceps suitable for use in corrosive environments. Capable of withstanding high temperatures, these forceps are ideal for applications that involve heat exposure.
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)