Merck
BAH379400000
SP Bel-Art® FEP Coated Steel Forceps
curved, sharp point PTFE-coated stainless steel forceps, L 3.875 in. (98 mm) , tip coating
About This Item
UNSPSC-Code:
41122800
NACRES:
NB.43
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Materialien
curved, sharp point PTFE-coated stainless steel forceps
Verpackung
pkg of 2 ea (48/case)
Hersteller/Markenname
Bel-Art H37940-0000
Länge
3.875 in. (98 mm) , tip coating
Allgemeine Beschreibung
The exceptionally fine tips of these forceps are coated with brown Fluo-Kem® PTFE FEP.
Leistungsmerkmale und Vorteile
  • Complies with FDA regulations covering direct contact with food.
  • Serrated, curved tips
Rechtliche Hinweise
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Fluo-kem is a registered trademark of SP Scienceware, a division of SP
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
Fragen
  1. Can these SP Bel-Art® FEP Coated Steel Forceps be autoclaved?
    1 Antwort
    1. This is a product of SP Bel-Art. The manufacturer states that this item is autoclavable.
      Hilfreich?