Skip to Content
Merck

Skip To

1217909

USP

Diphenhydramine citrate

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Benzhydramine citrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
200 mg
Please contact Customer Service for Availability
CZK 11,500.00

About This Item

Empirical Formula (Hill Notation):
C17H21NO · C6H8O7
CAS Number:
Molecular Weight:
447.48
MDL number:
UNSPSC Code:
41116107
EC Number:
289-432-0
NACRES:
NA.24

CZK 11,500.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

diphenhydramine

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

N(CCOC(c2ccccc2)c1ccccc1)(C)C.OC(CC(=O)O)(CC(=O)O)C(=O)O

InChI

1S/C17H21NO.C6H8O7/c1-18(2)13-14-19-17(15-9-5-3-6-10-15)16-11-7-4-8-12-16;7-3(8)1-6(13,5(11)12)2-4(9)10/h3-12,17H,13-14H2,1-2H3;13H,1-2H2,(H,7,8)(H,9,10)(H,11,12)

InChI key

SPCKHVPPRJWQRZ-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Diphenhydramine citrate USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Acetaminophen and Diphenhydramine Citrate Tablets
  • Diphenhydramine Citrate
  • Diphenhydramine Citrate and Ibuprofen Tablets
  • Orphenadrine Citrate

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
121800512180161218050
manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

format

neat

format

neat

format

neat

format

neat

API family

diphenhydramine

API family

diphenhydramine

API family

diphenhydramine

API family

diphenhydramine


Still not finding the right product?

Explore all of our products under Diphenhydramine citrate


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters