Skip to Content
Merck
All Photos(1)

Key Documents

YSAM

Sigma-Aldrich

Yeast

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
CZK 4,290.00

CZK 4,290.00


Please contact Customer Service for Availability


Select a Size

Change View
10 G
CZK 4,290.00

About This Item

UNSPSC Code:
12352204
NACRES:
NA.26

CZK 4,290.00


Please contact Customer Service for Availability

application(s)

food and beverages

storage temp.

−20°C

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service