Skip to Content
Merck
All Photos(1)

Key Documents

S8065

Sigma-Aldrich

Amyloid Precursor Protein α (304-612), Secreted human

>90% (SDS-PAGE), recombinant, expressed in E. coli (N-terminal histidine tagged), buffered aqueous solution

Synonym(s):

sAPPα (304-612)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
NACRES:
NA.32

recombinant

expressed in E. coli (N-terminal histidine tagged)

Assay

>90% (SDS-PAGE)

form

buffered aqueous solution

mol wt

apparent mol wt ~45 kDa by SDS-PAGE

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... APP(351)

Biochem/physiol Actions

Fragment of sAPPα designed to lack the N-terminal domain of the full length protein.
Secreted Amyloid Precursor Protein α (sAPPα) is a 612 amino acid protein produced from the ubiquitously expressed β-Amyloid precursor protein (βAPP), 695 isoform, by α-secretase cleavage. sAPPα (304-612) is a fragment of sAPPα designed to lack the N-terminal domain of the full length protein.

Physical form

0.2 μm filtered solution in phosphate buffered saline, pH 7.4.

Preparation Note

Expressed as a soluble protein and purified under non-denaturing conditions.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mattson, M.P.
The Journal of Neuroscience, 25, 439-439 (1994)
Greenfield, J.P.,
Frontiers in Bioscience, 5, 72-72 (2000)
L W Jin et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 14(9), 5461-5470 (1994-09-01)
Amyloid beta/A4 protein precursor (APP) is secreted into medium by most cultured cells and can function as an autocrine factor. To study the biological function of secreted forms of APP (sAPP) on neurons, we used a clonal CNS neuronal line
S W Barger et al.
Nature, 388(6645), 878-881 (1997-08-28)
A role for beta-amyloid precursor protein (beta-APP) in the development of Alzheimer's disease has been indicated by genetics, and many conditions in which beta-APP is raised have been associated with an increased risk of Alzheimer's disease or an Alzheimer's-like pathology.
Cell biology of the amyloid beta-protein precursor and the mechanism of Alzheimer's disease.
D J Selkoe
Annual review of cell biology, 10, 373-403 (1994-01-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service