Skip to Content
Merck
All Photos(1)

Key Documents

MAK324

Sigma-Aldrich

Acetylcholinesterase Inhibitor Screening Kit

sufficient for 100 colorimetric tests

Synonym(s):

AChE Inhibitor Assay Kit

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
CZK 10,400.00

CZK 10,400.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 KIT
CZK 10,400.00

About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

CZK 10,400.00


Please contact Customer Service for Availability

Request a Bulk Order

usage

sufficient for 100 colorimetric tests

detection method

colorimetric

relevant disease(s)

neurological disorders, Alzheimer′s disease

storage temp.

−20°C

General description

Acetylcholinesterase (AChE), also known as RBC cholinesterase, is found primarily in the blood and neural synapses. AChE catalyzes the hydrolysis of the neurotransmitter acetylcholine into choline and acetic acid, a reaction necessary to allow a cholinergic neuron to return to its resting state after activation. Inhibition of the enzyme leads to acetylcholine accumulation, hyperstimulation of nicotinic and muscarinic receptors, and disrupted neurotransmission. AChE inhibition is an important target for the management of Alzheimer′s disease and AChE inhibitors are the most common drugs used for its management. In addition to Alzheimer′s disease, AChE inhibitors have been useful in the diagnosis or treatment of diseases such as glaucoma, myasthenia gravis, bladder distention, and more. The Acetylcholinesterase Inhibitor Screening kit is based on an improved Ellman method, in which thiocholine produced by the action of acetylcholinesterase forms a yellow color with 5,5′-dithiobis(2-nitrobenzoic acid). The intensity of the product color, measured at 412 nm, is proportionate to the enzyme activity in the sample.

Application


  • In vitro assessment of selected Korean plants for antioxidant and antiacetylcholinesterase activities.: This research evaluates the antioxidant and anti-acetylcholinesterase activities of various Korean plants using in vitro screening, showcasing the screening kit′s application in natural product research (Lee et al., 2017).

The Acetylcholinesterase Inhibitor Screening Kit may be used for:
  • Drug Discovery
  • Neurological Disease Research
  • Alzheimer′s Disease Research

Features and Benefits

Efficient Inhibitor Screening: Screen for AChE inhibitors with only 5 μL of test compound and a short 15 minute reaction time, offering a streamlined and effortless procedure.

Compatibility with High-Throughput Systems: Easily incorporate our kit into high-throughput handling systems, ensuring smooth and accurate processing, enhancing efficiency in your laboratory workflow.

Suitability

Suitable for inhibitor screening and evaluation of acetylcholinesterase inhibitors.

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dawid Maliszewski et al.
Molecules (Basel, Switzerland), 26(13) (2021-07-03)
A series of new analogs of nitrogen mustards (4a-4h) containing the 1,3,5-triazine ring substituted with dipeptide residue were synthesized and evaluated for the inhibition of both acetylcholinesterase (AChE) and β-secretase (BACE1) enzymes. The AChE inhibitory activity studies were carried out

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service