Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
50 μg | Please contact Customer Service for Availability | CZK 16,400.00 |
About This Item
recombinant
expressed in HEK 293 cells
Quality Segment
assay
≥98% (SDS-PAGE)
form
liquid
potency
Supports transfection of Simplicon-based TagGFP RNA
mol wt
50 kDa (glycosylated)
technique(s)
cell culture | mammalian: suitable
suitability
endotoxin tested
shipped in
dry ice
storage temp.
−70°C
Gene Information
vaccinia virus ... B18R(3707576)
General description
Application
Biochem/physiol Actions
Physical form
1 of 1
This Item | |||
|---|---|---|---|
| assay ≥98% (SDS-PAGE) | assay >98% (SDS-PAGE) | assay ≥98% (SDS-PAGE) | assay ≥95% (SDS-PAGE) |
| recombinant expressed in HEK 293 cells | recombinant - | recombinant expressed in HEK 293 cells | recombinant expressed in HEK 293 cells |
| technique(s) cell culture | mammalian: suitable | technique(s) cell culture | stem cell: suitable | technique(s) cell culture | mammalian: suitable | technique(s) cell culture | mammalian: suitable |
| suitability endotoxin tested | suitability - | suitability endotoxin tested | suitability - |
| form liquid | form liquid | form lyophilized powder | form lyophilized powder |
| mol wt 50 kDa (glycosylated) | mol wt Mw 38,389 Da | mol wt 16 kDa (glycosylated) | mol wt monomer 18 kDa (glycosylated) |
Storage Class
10 - Combustible liquids
wgk
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



