Skip to Content
Merck

Skip To

E1500

Elastin-orcein

elastase substrate

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
12352204
NACRES:
NA.32
MDL number:


form

powder

storage temp.

2-8°C

General description

Substrate for the estimation of elastase.

Preparation Note

Prepared from E1625.

Other Notes

Elastin impregnated with orcein.


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Articles

Elastase application index for understanding leukocyte elastase, a 29KDa serine endoprotease.


Yuko Ohara-Nemoto et al.
FEMS microbiology letters, 367(24) (2020-12-19)
Multiple dipeptidyl-peptidases (DPPs) are present in the periplasmic space of Porphyromonas gingivalis, an asaccharolytic periodontopathic bacterium. Dipeptides produced by DPPs are presumed to be transported into the bacterial cells and metabolized to generate energy and cellular components. The present study
Hee Chan Yoo et al.
Cell metabolism, 31(2), 267-283 (2019-12-24)
Glutamine is an essential nutrient that regulates energy production, redox homeostasis, and signaling in cancer cells. Despite the importance of glutamine in mitochondrial metabolism, the mitochondrial glutamine transporter has long been unknown. Here, we show that the SLC1A5 variant plays



Global Trade Item Number

SKUGTIN
E1500-5G04061832923895

Questions

Reviews

No rating value

Active Filters