Skip to Content
Merck

Skip To

C6482

Caspase 5 human

recombinant, expressed in E. coli, 5,000 units/mg protein

Synonym(s):

ICE(rel)III, TY

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
100 units
Please contact Customer Service for Availability
CZK 19,600.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32
MDL number:
Specific activity:
5,000 units/mg protein
Recombinant:
expressed in E. coli

CZK 19,600.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


recombinant

expressed in E. coli

Quality Segment

form

powder

specific activity

5,000 units/mg protein

mol wt

30 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... CASP5(838)

Biochem/physiol Actions

Useful in screening caspase inhibitors, studying enzyme kinetics and regulation, determining target substrates, as well as serving as positive controls in caspase activity assays and Western blot analysis.

Physical form

Powder contains 0.052% ammonium sulfate, 0.158% Tris−HCl, and 0.76% sodium chloride.

Other Notes

One unit will hydrolyze 1 nmol of the caspase substrate WEHD-pNA to WEHD and p-nitroaniline per hour at pH 7.2 at 37 °C.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PRS3455C6357C1224
Gene Information

human ... CASP5(838)

Gene Information

human ... CASP5(838)

Gene Information

human ... CASP4(837)

Gene Information

human ... CASP3(836)

specific activity

5,000 units/mg protein

specific activity

-

specific activity

5,000 units/mg protein

specific activity

≥1.0 units/mg protein

form

powder

form

buffered aqueous solution

form

solution

form

buffered aqueous glycerol solution

recombinant

expressed in E. coli

recombinant

-

recombinant

expressed in E. coli

recombinant

expressed in E. coli (C-terminal histidine-tagged)

UniProt accession no.

P51878

UniProt accession no.

P51878

UniProt accession no.

P49662

UniProt accession no.

P42574

storage temp.

−70°C

storage temp.

−20°C

storage temp.

−70°C

storage temp.

−70°C


Storage Class

10 - Combustible liquids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content






Global Trade Item Number

SKUGTIN
C6482-100UN04061833629192
C6482-25UN04061833629208

Questions

Reviews

No rating value

Active Filters