Skip to Content
Merck

Skip To

C0788

Z-Leu-Leu-Glu β-naphthylamide

≥98%, powder

Synonym(s):

Cbz-LLE β-naphthylamine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
50 mg
Please contact Customer Service for Availability
CZK 7,680.00

About This Item

Empirical Formula (Hill Notation):
C35H44N4O7
CAS Number:
Molecular Weight:
632.75
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352209
MDL number:
Assay:
≥98%
Form:
powder
Quality level:

CZK 7,680.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

assay

≥98%

form

powder

color

white

solubility

THF: 25 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

CC(C)CC(NC(=O)OCc1ccccc1)C(=O)NC(CC(C)C)C(=O)NC(CCC(O)=O)C(=O)Nc2ccc3ccccc3c2

InChI

1S/C35H44N4O7/c1-22(2)18-29(38-34(44)30(19-23(3)4)39-35(45)46-21-24-10-6-5-7-11-24)33(43)37-28(16-17-31(40)41)32(42)36-27-15-14-25-12-8-9-13-26(25)20-27/h5-15,20,22-23,28-30H,16-19,21H2,1-4H3,(H,36,42)(H,37,43)(H,38,44)(H,39,45)(H,40,41)

InChI key

MIYHULQEZFNUGE-UHFFFAOYSA-N

General description

Substrate for the glutamylpeptidyl-peptide hydrolase activity of the 20S proteasome (multicatalytic proteinase complex).

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
L1635SML1836C0483
assay

≥98%

assay

≥98% (TLC)

assay

≥98% (HPLC)

assay

≥95%

form

powder

form

powder

form

powder

form

solid

Quality Level

200

Quality Level

200

Quality Level

100

Quality Level

200

storage temp.

−20°C

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

−20°C

solubility

THF: 25 mg/mL, clear, colorless

solubility

H2O: insoluble

solubility

0.1 M HCl: 2 mg/mL, clear (warmed)

solubility

DMSO: soluble

color

white

color

-

color

white to beige

color

-


pictograms

Health hazard

signalword

Warning

hcodes

pcodes

Hazard Classifications

Carc. 2

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



H Djaballah et al.
Biochemistry, 31(16), 4133-4141 (1992-04-28)
The multicatalytic proteinase (MCP) complex or proteasome is a major nonlysosomal proteinase of eukaryotic cells. The proteinase can cleave peptide bonds on the carboxyl side of hydrophobic, basic, or acidic amino acid residues. These activities have been referred to as
C Realini et al.
The Journal of biological chemistry, 272(41), 25483-25492 (1997-11-05)
Full-length cDNAs for three human proteasome activator subunits, called REGalpha, REGbeta, and REGgamma, have been expressed in Escherichia coli, and the purified recombinant proteins have been characterized. Recombinant alpha or gamma subunits form heptameric species; recombinant beta subunits are found
C Cardozo
Enzyme & protein, 47(4-6), 296-305 (1993-01-01)
The multicatalytic proteinase complex (MPC), also called the proteasome, is a ubiquitous particle (19S) that is required for life. It is found in the cytoplasm and nucleus of all eukaryotic cells where it degrades selected cytosolic and nuclear proteins. It



Global Trade Item Number

SKUGTIN
C0788-50MG04061832269801
C0788-10MG04061833260654
C0788-25MG04061833260661

Questions

Reviews

No rating value

Active Filters