Skip to Content
Merck

Skip To

BP484

Clonazepam

British Pharmacopoeia (BP) Reference Standard

Synonym(s):

5-(2-Chlorophenyl)-7-nitro-3H-1,4-benzodiazepin-2(1H)-one

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 mg
Please contact Customer Service for Availability
CZK 6,360.00

About This Item

Empirical Formula (Hill Notation):
C15H10ClN3O3
CAS Number:
Molecular Weight:
315.71
UNSPSC Code:
41116107
NACRES:
NA.24
MDL number:

CZK 6,360.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

API family

clonazepam

form

solid

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

BP

drug control

kontrollierte Droge in Deutschland, psicótropo (Spain); Decreto Lei 15/93: Tabela IV (Portugal); Pszichotróp anyag / Psychotropic Substance (Hungary), 78/2022. (XII. 28.) BM rendelet

application(s)

pharmaceutical
pharmaceutical small molecule

format

neat

storage temp.

2-8°C

SMILES string

Clc1ccccc1C2=NCC(=O)Nc3ccc(cc23)N(=O)=O

InChI

1S/C15H10ClN3O3/c16-12-4-2-1-3-10(12)15-11-7-9(19(21)22)5-6-13(11)18-14(20)8-17-15/h1-7H,8H2,(H,18,20)

InChI key

DGBIGWXXNGSACT-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Clonazepam BP Reference standard, intended for use in laboratory tests only as specifically prescribed in the British Pharmacopoeia.

Also used in monographs such as:
  • Clonazepam Oral Suspension
  • Clobazam Oral Suspension
  • Nitrazepam Oral Suspension
  • Clonazepam Tablets
  • Nitrazepam Tablets
  • Clonazepam Injection
  • Packaging

    Unit quantity: 100 mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity please visit British Pharmacopoeia

    Other Notes

    Sales restrictions may apply.

    Legal Information

    German
    Dieses Produkt fällt unter das Betäubungsmittelgesetz (BtMG). Für eine Bestellung dieses Produktes ist eine Erlaubnis nach § 3 BtMG zwingend erforderlich, es sei denn, es greift eine Ausnahme von der Erlaubnispflicht nach § 4 oder § 26 BtMG.

    English
    This product is subject to the German Narcotics Act. A permit under Section 3 of the German Narcotics Act is mandatory for ordering this product unless an exemption from the permit requirement under Section 4 or Section 26 of the German Narcotics Act applies.

    Compare Similar Items

    View Full Comparison

    Show Differences

    1 of 1

    This Item
    BP507BP434BP840
    Clonazepam British Pharmacopoeia (BP) Reference Standard

    BP484

    Clonazepam

    Clobazam British Pharmacopoeia (BP) Reference Standard

    BP507

    Clobazam

    Lorazepam British Pharmacopoeia (BP) Reference Standard

    BP434

    Lorazepam

    grade

    pharmaceutical primary standard

    grade

    pharmaceutical primary standard

    grade

    pharmaceutical primary standard

    grade

    pharmaceutical primary standard

    manufacturer/tradename

    BP

    manufacturer/tradename

    BP

    manufacturer/tradename

    BP

    manufacturer/tradename

    BP

    form

    solid

    form

    solid

    form

    solid

    form

    solid

    shelf life

    limited shelf life, expiry date on the label

    shelf life

    limited shelf life, expiry date on the label

    shelf life

    limited shelf life, expiry date on the label

    shelf life

    limited shelf life, expiry date on the label

    format

    neat

    format

    neat

    format

    neat

    format

    neat

    application(s)

    pharmaceutical
    pharmaceutical small molecule

    application(s)

    pharmaceutical
    pharmaceutical small molecule

    application(s)

    pharmaceutical
    pharmaceutical small molecule

    application(s)

    pharmaceutical
    pharmaceutical small molecule


    Still not finding the right product?

    Explore all of our products under Clonazepam


    pictograms

    Exclamation mark

    signalword

    Warning

    hcodes

    Hazard Classifications

    Aquatic Chronic 3 - STOT SE 3

    target_organs

    Central nervous system

    Storage Class

    11 - Combustible Solids

    wgk

    WGK 1

    flash_point_f

    Not applicable

    flash_point_c

    Not applicable



    Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

    If you need assistance, please contact Customer Support

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library



    Questions

    Reviews

    No rating value

    Active Filters