Skip to Content
Merck

Skip To

89503

Ammonium Nitrogen Standard for IC

1000 mg/L N as NH4+ in water (nominal concentration), TraceCERT®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
100 mL
Please contact Customer Service for Availability
CZK 1,150.00

About This Item

NACRES:
NA.24
UNSPSC Code:
41116107

CZK 1,150.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

product line

TraceCERT®

grade

certified reference material, TraceCERT®

concentration

1000 mg/L N as NH4+ in water (nominal concentration)

technique(s)

ion chromatography: suitable

application(s)

environmental
food and beverages

format

single component solution

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to SI unit kg and measured against primary material from an NMI, e.g. NIST.
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Analysis Note

1000 mg/L N as NH4+ in water, prepared with high purity NH4Cl and water

Other Notes

Visit the IC Portal to learn more

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
597555363836427
application(s)

environmental
food and beverages

application(s)

environmental
food and beverages

application(s)

environmental
food and beverages

application(s)

environmental
food and beverages

technique(s)

ion chromatography: suitable

technique(s)

ion chromatography: suitable

technique(s)

ion chromatography: suitable

technique(s)

ion chromatography: suitable

format

single component solution

format

single component solution

format

single component solution

format

single component solution

Quality Level

300

Quality Level

300

Quality Level

300

Quality Level

300

grade

certified reference material, TraceCERT®

grade

certified reference material, TraceCERT®

grade

certified reference material, TraceCERT®

grade

certified reference material, TraceCERT®

concentration

1000 mg/L N as NH4+ in water (nominal concentration)

concentration

1000 mg/L NH4+ in water (nominal concentration)

concentration

1000 mg/L N as nitrate in water (nominal concentration)

concentration

1000 mg/L N as nitrite in water (pH ~11 nominal concentration)


Storage Class

12 - Non Combustible Liquids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

12125-02-9

CAS No.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Articles

Comprehensive water analysis with Spectroquant® systems, test kits, and reference materials for accurate, reliable, and compliant results.

Complete wastewater testing solutions for every treatment stage with instruments, test kits, and standards to ensure compliant, reliable water analysis.





Global Trade Item Number

SKUGTIN
89503-100ML04061837712722

Questions

  1. The way this is named is super confusing. Normally, I would see NH4+ standard written "as N" meaning that only the nitrogen is considered toward the concentration. This seems to indicate that the NH4+ molecular weight is used to calculate the conc!?!

    1 answer
    1. This product is not an ammonium standard, but a nitrogen standard (in the form of ammonium). If written, 'NH4+ standard as N', that would imply that the product is an ammonium standard, which this is not; it is an ammonium *nitrogen* standard.

      Helpful?

Reviews

No rating value

Active Filters