Skip to Content
Merck

Skip To

S7108

ApopTag Stop/Wash Buffer

ApopTag Stop/Wash Buffer is intended for use in labeling the 3′-OH ends of fragmented DNA during apoptotic cell detection.

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
20 mL

Estimated to ship onMay 12, 2026fromAreál Kühne+Nagel spol. s r.o.

CZK 4,760.00

About This Item

UNSPSC Code:
12161503
NACRES:
NA.32
eCl@ss:
32161000

CZK 4,760.00


Estimated to ship onMay 12, 2026Details


Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

manufacturer/tradename

ApopTag, Chemicon®

technique(s)

activity assay: suitable (apoptosis)

shipped in

dry ice

General description

ApopTag Stop/Wash Buffer is intended for use in labeling the 3′-OH ends of fragmented DNA during apoptotic cell detection. Qualified for use with Apoptag In Situ Apoptosis Detection Kits (Catalog Numbers: S7100, S7101, S7111 and S7165).

Materials Provided

Each bottle contains 20 ml of ApopTag Stop/Wash Buffer.



ApopTag is a registered trademark of Serologicals Company.

Application

Instructions for Use

Prepare Working Strength Stop/Wash Buffer by adding:

1 ml ApopTag Stop/Wash Buffer

34 ml dH2O

35 ml Total



This amount is sufficient to treat 5 slides in a coplin jar. Working Strength Stop/Wash Buffer may be prepared in advance and stored in a glass or plastic container at 4°C for up to one (1) year. Use a fresh aliquot for each experiment.



Working Strength Stop/Wash Buffer is ready for use at the "APPLY STOP/WASH BUFFER" protocol step in The Complete ApopTag Manual.



NOTE: Agitate for 15 seconds and incubate for 10 min. at room temperature to avoid precipitation.

Preparation Note

Recommended Storage is -15°C to -25°C.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
S7106S7105S7100
technique(s)

activity assay: suitable (apoptosis)

technique(s)

activity assay: suitable (apoptosis)

technique(s)

activity assay: suitable (apoptosis)

technique(s)

-

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

manufacturer/tradename

ApopTag, Chemicon®

manufacturer/tradename

ApopTag, Chemicon®

manufacturer/tradename

ApopTag, Chemicon®

manufacturer/tradename

ApopTag, Chemicon®

shipped in

dry ice

shipped in

dry ice

shipped in

dry ice

shipped in

dry ice


pictograms

Health hazardCorrosion

signalword

Danger

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A - STOT RE 2 Inhalation

target_organs

Respiratory Tract

Storage Class

8B - Non-combustible corrosive hazardous materials

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
S710804053252578083

Questions

Reviews

No rating value

Active Filters