Skip to Content
Merck
All Photos(1)

Key Documents

S7106

Sigma-Aldrich

ApopTag Equilibration Buffer

ApopTag Equilibration Buffer is intended for use in labeling the 3′-OH ends of fragmented DNA during apoptotic cell detection.

Sign Into View Organizational & Contract Pricing

Select a Size

15 ML
CZK 4,640.00

CZK 4,640.00


Estimated to ship onApril 14, 2025Details


Request a Bulk Order

Select a Size

Change View
15 ML
CZK 4,640.00

About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.84

CZK 4,640.00


Estimated to ship onApril 14, 2025Details


Request a Bulk Order

Quality Level

manufacturer/tradename

ApopTag
Chemicon®

technique(s)

activity assay: suitable (apoptosis)

shipped in

dry ice

General description

ApopTag Equilibration Buffer is intended for use in labeling the 3′-OH ends of fragmented DNA during apoptotic cell detection. Qualified for use with Apoptag In Situ Apoptosis Detection Kits (Catalog #s: S7100, S7101, S7110, S7111, S7160 and S7165).



Materials Provided

Each bottle contains 15 ml of ApopTag Equilibration Buffer.



ApopTag is a registered trademark of Serologicals Company.

Application

Instructions for Use

ApopTag Equilibration Buffer is ready for use at the "APPLY EQUILIBRATION BUFFER" step in The Complete ApopTag Manual.

Warnings and Precautions

ApopTag Equilibration Buffer contains potassium cacodylate (dimethylarsinic acid) as a buffer. This material is harmful if swallowed; avoid contact with skin and eyes (wear gloves, glasses); wash areas of contact immediately.

Storage and Stability

Recommended Storage -15°C to -25°C

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Eye Irrit. 2 - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Sens. 1

WGK

WGK 3


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service