Skip to Content
Merck

Skip To

8.07501

2-Pentanol

for synthesis

Synonym(s):

2-Pentanol, Methyl propyl carbinol, sec-n-Amyl alcohol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 mL
Please contact Customer Service for Availability
CZK 876.00
500 mL
Please contact Customer Service for Availability
CZK 2,130.00

About This Item

Linear Formula:
CH3CH2CH2CH(OH)CH3
CAS Number:
Molecular Weight:
88.15
UNSPSC Code:
12352104
EC Index Number:
227-907-6
NACRES:
NA.22
MDL number:
Form:
liquid

CZK 876.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

3 hPa ( 20 °C)

Quality Segment

form

liquid

autoignition temp.

330 °C

potency

1470 mg/kg LD50, oral (Rat), 5250 mg/kg LD50, skin (Rabbit)

expl. lim.

1.2-10.5 % (v/v)

bp

119 °C/1013 hPa

mp

-50 °C

transition temp

flash point 41 °C

density

0.81 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

OC(CCC)C

InChI

1S/C5H12O/c1-3-4-5(2)6/h5-6H,3-4H2,1-2H3

InChI key

JYVLIDXNZAXMDK-UHFFFAOYSA-N


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
8.075028.058048.06031
Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

density

0.81 g/cm3 at 20 °C

density

0.82 g/cm3 at 20 °C

density

0.81 g/cm3 at 20 °C

density

0.82 g/cm3 at 20 °C

bp

119 °C/1013 hPa

bp

115-116 °C/1013 hPa

bp

130-133 °C/1013 hPa

bp

-

form

liquid

form

liquid

form

liquid

form

liquid

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

2-30°C

mp

-50 °C

mp

-8 °C

mp

-60 °C

mp

-70 °C


Still not finding the right product?

Explore all of our products under 2-Pentanol


pictograms

FlameExclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

93.2 °F - closed cup

flash_point_c

34 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters