Skip to Content
Merck

Skip To

8.06031

2-Methyl-1-butanol

for synthesis

Synonym(s):

2-Methyl-1-butanol, 2-Methylbutyl alcohol, sec-Butyl carbinol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 L
Please contact Customer Service for Availability
CZK 1,950.00

About This Item

Linear Formula:
CH3CH2CH(CH3)CH2OH
CAS Number:
Molecular Weight:
88.15
UNSPSC Code:
12352104
EC Index Number:
205-289-9
NACRES:
NA.22
MDL number:
Assay:
≥98.0% (GC)
Form:
liquid

CZK 1,950.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

4.3 hPa ( 20 °C)

Quality Segment

assay

≥98.0% (GC)

form

liquid

autoignition temp.

340 °C

potency

4170 mg/kg LD50, oral (Rat), 2900 mg/kg LD50, skin (Rabbit)

expl. lim.

1.2-10.3 % (v/v)

pH

7 ( in H2O)

mp

-70 °C

transition temp

flash point 42.5 °C

solubility

36 g/L

density

0.82 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

OCC(CC)C

InChI

1S/C5H12O/c1-3-5(2)4-6/h5-6H,3-4H2,1-2H3

InChI key

QPRQEDXDYOZYLA-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.817 - 0.819
Identity (IR): passes test

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
8.222638.2092565990
assay

≥98.0% (GC)

assay

≥99% (GC)

assay

≥98% (GC)

assay

≥98.0% (GC)

solubility

36 g/L

solubility

125 g/L

solubility

-

solubility

-

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

form

liquid

form

liquid

form

oil

form

-

density

0.82 g/cm3 at 20 °C

density

0.81 g/cm3 at 20 °C

density

0.82 g/cm3 at 20 °C

density

0.819 g/mL at 20 °C (lit.)

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

-


Still not finding the right product?

Explore all of our products under 2-Methyl-1-butanol


signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk

WGK 1

flash_point_f

108.5 °F - closed cup

flash_point_c

42.5 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters