Skip to Content
Merck

Skip To

71748-M

Floating Rack, Mega

Synonym(s):

Floating Rack for D-Tube Dialyzers

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
10 ea

Available to ship TODAYfromAreál Kühne+Nagel spol. s r.o.

CZK 1,340.00

About This Item

NACRES:
NB.24
UNSPSC Code:
41104900
Manufacturer/tradename:
Novagen®

CZK 1,340.00


Available to ship TODAYDetails


Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


manufacturer/tradename

Novagen®

storage condition

OK to freeze

storage temp.

10-30°C

General description

The Floating Rack is designed to hold D-Tube Dialyzers in an exchange buffer. Each Floating Rack, Mini can hold up to nine D-Tube Dialyzer, Mini devices. Each Floating Rack, Midi can hold up to four D-Tube Dialyzer, Midi units. Each Floating Rack, Maxi can hold up to seven D-Tube Dialyzer, Maxi devices. Each Floating Rack, Mega can hold one D-Tube Dialyzer, Mega unit. Each package contains 10 Floating Racks.

Features and Benefits

  • Easy-to-handle dialyzers for buffer exchange and removal of urea, detergents
  • Ideal for electroelution of proteins, protein-DNA complexes, oligonucleotides, DNA, and RNA from polyacrylamide and agarose gels
  • One-step procedure that does not require syringes or any special equipment
  • Sample volume recovery >97%
  • Protease, RNAse, DNAse, and PCR product free

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
71514-M71512-MR7901
manufacturer/tradename

Novagen®

manufacturer/tradename

Novagen®

manufacturer/tradename

Novagen®

manufacturer/tradename

Heathrow Scientific HS2166

storage temp.

10-30°C

storage temp.

10-30°C

storage temp.

10-30°C

storage temp.

-

storage condition

OK to freeze

storage condition

OK to freeze

storage condition

OK to freeze

storage condition

-


Storage Class

10-13 - German Storage Class 10 to 13



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library





Global Trade Item Number

SKUGTIN
71748-304055977266993

Questions

Reviews

No rating value

Active Filters