Skip to Content
Merck

Skip To

69725-M

λDE3 Lysogenization Kit plus pLysS & pLysE - Novagen

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

NACRES:
NA.51
UNSPSC Code:
41106200
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


manufacturer/tradename

Novagen®

Quality Level

storage condition

OK to freeze, avoid repeated freeze/thaw cycles

storage temp.

−70°C

General description

The λDE3 Lysogenization Kit is designed for site-specific integration of λDE3 prophage into an E. coli host cell chromosome, such that the lysogenized host can be used to express target genes cloned in pET vectors. λDE3 is a recombinant phage carrying the cloned gene for T7 RNA polymerase under lacUV5 control. Lysogens are prepared by co-infection with the three provided phages, and can be verified by plating the T7 Tester Phage (a T7 RNA polymerase deletion mutant). This phage is unable to make a plaque on cells that lack T7 RNA polymerase, but makes normal plaques on λDE3 lysogens in the presence of IPTG.The T7 RNA Polymerase Monoclonal Antibody can be used to verify the presence of T7 RNA polymerase in the host strain by Western blot analysis.

This product contains genetically modified organisms (GMO). Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively. This legislation obliges us to request certain information about you and the establishment where the GMOs are being handled. Click here for Enduser Declaration (EUD) Form.

Other Notes

•200 µlλDE3 Phage Lysate

•200 µlHelper Phage Lysate

•200 µlSelection Phage Lysate

•50 µlT7 Tester Phage Lysate

•200 µlHMS174(DE3) glycerol stock

•1 µgpLysS DNA

•1 µgpLysE DNA

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Multiple Toxicity Values, refer to MSDS (O)


Storage Class

10 - Combustible liquids

wgk

WGK 2



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library






Global Trade Item Number

SKUGTIN
69725-304055977258981

Questions

Reviews

No rating value

Active Filters