Skip to Content
Merck
All Photos(1)

Key Documents

5.09911

Sigma-Aldrich

RNA Polymerase I Inhibitor II, BMH-21

Synonym(s):

RNA Polymerase I Inhibitor II, BMH-21, BMH21, rRNA Transcription Inhibitor II, N-(2-(Dimethylamino)ethyl)-12-oxo-12H-benzo[g]pyrido[2,1-b]quinazoline-4-carboxamide, N-(2-(Dimethylamino)ethyl)-12-oxo-12H-benzo[g]pyrido[2,1-b]quinazoline-4-carboxamide, BMH21, rRNA Transcription Inhibitor II

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H20N4O2
CAS Number:
Molecular Weight:
360.41
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Assay

≥98% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

orange-yellow

solubility

DMSO: 1 mg/mL
5% acetic acid: 25 mg/mL

storage temp.

2-8°C

SMILES string

[n]21c(nc3c([c]2=O)cc4c(c3)cccc4)c(ccc1)C(=O)NCCN(C)C

InChI key

BXYDVWIAGDJBEC-UHFFFAOYSA-N

General description

A cell-permeable, non-toxic, benzopyridoquinazoline-carboxamide compound that blocks the growth and viability of a variety of cancer cell lines (GI50 = 160 nM in NCI-60 cells), but does not significantly affect normal cells (~90-fold therapeutic window). Preferentially binds to the GC-rich DNA and inhibits RNA polymerase I (Pol I) transcription. Although it intercalates with DNA, it does not activate the DNA damage response. Induces a proteasome-dependent destruction of large Pol I catalytic subunit RPA194, independent of TP53 genetic status of cancer cells, leading to disassembly of Pol I holocomplex from the rDNA. However, it does not affect Rpb1, the large subunit of RNA polymerase II and Pol I preinitiation complex proteins. Effectively reduces the growth of A375 and HCT116 xenografts in athymic NCr nu/nu mice (50 mg/kg, i.p, 2 weeks).

Please note that the molecular weight for this compound is batch-specific due to variable water content.
A cell-permeable, reversible DNA-intercalating benzopyridoquinazoline-carboxamide that preferentially targets GC-rich sequence, notably that of rDNA (162% of average human genome GC content), and effectively inhibits RNA polymerase I- (Pol I) mediated rDNA transcription both in cell-free assays and in cultures (IC50/ICmax = 60 nM/≤1 M against 2-h de novo 47S transcription in A375 cells) without affecting the maturation/processing of 47S into 32S and 18S rRNA. Time-dependent studies in 1 M BMH-21-treated A375 cells reveal fast inhibition of nuclear RNA synthesis (FUrd incorporation) within 15 min, followed by progressive altered localization of nucleolar proteins (starting in <20 min), including Pol I subunit RPA194 capping structure formation, indicative of stalled Pol I complex, and eventual proteasome-mediated RPA194 degradation (>1 h). Although BMH-21 activates p53 independent of DNA damage-sensing ATM pathway signaling in A375 cultures, wt p53 is not a prerequisite for BMH-21 anticancer activity (Av GI50 = 110 nM/wt and 205 nM/mutant among NCI60 cancer panel). Intraperitoneal injection is demonstrated to be efficacious in suppressing A375 (25 & 50 mg/kg/d, 6d/wk) and HCT-116 (50 mg/kg/d, 7 d/wk) tumor growth in mice in vivo. MG-132 (Cat. Nos. 474790, 474787, 474788, and 474791) effectively prevents BMH-21-induced RPA194 degradation without restoring stalled rRNA synthesis.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
rDNA
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Peltonen, K., et al. 2014. Cancer Cell25, 77.

Peltonen, K., et al. 2010. PLoS One5, e12996.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service