Skip to Content
Merck

Skip To

476475

PKC Inhibitor, EGF-R Fragment 651-658, Myristoylated

The PKC Inhibitor, EGF-R Fragment 651-658, Myristoylated controls the biological activity of PKC. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

PKC Inhibitor, EGF-R Fragment 651-658, Myristoylated, Myr-N-RKRTLRRL-OH, Myristoylated EGF-R Fragment (651-658), Protein Kinase C Inhibitor, Protein Kinase C Inhibitor, EGF-R Fragment 651-658, Myristoylated, Myristoylated EGF-R Fragment (651-658), Protein Kinase C Inhibitor, Myr-N-RKRTLRRL-OH, Protein Kinase C Inhibitor, EGF-R Fragment 651-658, Myristoylated

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
500 μg

Estimated to ship onJune 03, 2026fromAreál Kühne+Nagel spol. s r.o.

CZK 5,400.00

About This Item

Empirical Formula (Hill Notation):
C60H117N21O11
Molecular Weight:
1308.71
NACRES:
NA.77
UNSPSC Code:
12352202
Assay:
≥97% (HPLC)
Form:
lyophilized solid
Storage condition:
OK to freeze, desiccated

CZK 5,400.00


Estimated to ship onJune 03, 2026Details


Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥97% (HPLC)

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, desiccated

solubility

water: 5 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

Epidermal growth-factor receptor (EGF-R) conserved sequence which is identical to v-erbB (95-102). An N-terminal myristoylated membrane-permeable, reversible, and substrate competitive inhibitor that inhibits protein kinase C (IC50 = 5 µM) in intact cells.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
PKC
Product does not compete with ATP.
Reversible: yes
Target IC50: 5 µM against protein kinase C

Packaging

Yes

Physical form

Supplied as a trifluoroacetate salt.

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 2 months at -20°C.

Other Notes

Myr-N-Arg-Lys-Arg-Thr-Leu-Arg-Arg-Leu-OH
Ward, N.E., and O’Brian, C.A. 1993. Biochemistry32, 11903.
O’Brian, C.A., et al. 1991. Invest. New Drugs 9, 169.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
539604539636476485
form

lyophilized solid

form

lyophilized solid

form

lyophilized solid

form

lyophilized

assay

≥97% (HPLC)

assay

≥95% (HPLC)

assay

≥95% (HPLC)

assay

≥95% (HPLC)

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

solubility

water: 5 mg/mL

solubility

water: 1 mg/mL

solubility

water: 2 mg/mL

solubility

water: 1 mg/mL


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters