Skip to Content
Merck

Skip To

344906

FURA 2/AM in Solution

Synonym(s):

FURA 2/AM in Solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
1 mL
Check Cart for Availability
CZK 7,370.00

About This Item

Empirical Formula (Hill Notation):
C44H47N3O24
CAS Number:
Molecular Weight:
1001.85
UNSPSC Code:
12171500
NACRES:
NA.47
MDL number:

CZK 7,370.00


Check Cart for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥95% (HPLC)

Quality Segment

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, protect from light

fluorescence

λex 335 nm (high), λex 362 nm (low), λem 505 nm (high), λem 512 nm (low)

shipped in

ambient

storage temp.

−20°C

SMILES string

N(CC(=O)OCOC(=O)C)(CC(=O)OCOC(=O)C)c1c(cc(cc1)C)OCCOc2c3c([o]c(c3)c4ncc([o]4)C(=O)OCOC(=O)C)ccc2N(CC(=O)OCOC(=O)C)CC(=O)OCOC(=O)C

InChI

1S/C44H47N3O24/c1-25-7-8-32(46(16-38(53)65-20-60-26(2)48)17-39(54)66-21-61-27(3)49)35(13-25)58-11-12-59-42-31-14-36(43-45-15-37(71-43)44(57)69-24-64-30(6)52)70-34(31)10-9-33(42)47(18-40(55)67-22-62-28(4)50)19-41(56)68-23-63-29(5)51/h7-10,13-15H,11-12,16-24H2,1-6H3

InChI key

UTBYGIDPBQOHNT-UHFFFAOYSA-N

General description

Cell-permeable ester form of FURA 2 (Cat. No. 344900). The active dye is rapidly released in the cell by cytoplasmic esterases. Fluorescent calcium chelator (Kd = 224 nM) that offers 30X greater fluorescence intensity that QUIN 2 (Cat. No. 551826). This probe has been used as a probe in many cellular systems, including cardiac myocytes, vascular smooth muscle cells, and medullary collecting tubules. Spectral properties in the presence of low calcium concentrations (excitation max.: 362 nm, emission max.: 512 nm) differ from those during high calcium conditions (excitation max.: 335 nm, emission max.: 505 nm).

Biochem/physiol Actions

Cell permeable: no
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Physical form

A 1 mM solution of FURA 2/AM (Cat. No. 344905) in anhydrous DMSO.

Preparation Note

Following initial thaw, aliquot and freeze (-20°C). Store frozen under an inert atmosphere.

Other Notes

Davis, M.J., et al. 1992. Am. J. Physiol. 263, H1292.
Rao, G.H., et al. 1992. Biochem. Med. Metab. Biol.47, 226.
Schwartz, Y., et al. 1992. Endocrinology131, 772.
Ward, C.A., and Moffat, M.P. 1992. J. Mol. Cell. Cardiol.24, 937.
Janconi, M.E., et al. 1990. J. Cell Biol.110, 1555.
Tsien, R.Y. 1980. Biochemistry19, 2396.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Irritant (B)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
F088834490547989
fluorescence

λex 335 nm (high), λem 505 nm (high), λex 362 nm (low), λem 512 nm (low)

fluorescence

-

fluorescence

λex 335 nm (high), λex 362 nm (low), λem 505 nm (high), λem 512 nm (low)

fluorescence

λex 355 nm; λem 495 nm in 0.1 M Tris pH 8.0, esterase; 10 mM Ca2+

form

liquid

form

powder

form

solid

form

solid

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

shipped in

ambient

shipped in

-

shipped in

ambient

shipped in

-

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

assay

≥95% (HPLC)

assay

≥95% (HPLC)

assay

≥95% (HPLC)

assay

≥95.0% (HPLC)


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

188.6 °F - closed cup - (Dimethylsulfoxide)

flash_point_c

87 °C - closed cup - (Dimethylsulfoxide)



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Rolando Carrisoza-Gaytan et al.
JCI insight, 5(8) (2020-04-08)
BK channels are expressed in intercalated cells (ICs) and principal cells (PCs) in the cortical collecting duct (CCD) of the mammalian kidney and have been proposed to be responsible for flow-induced K+ secretion (FIKS) and K+ adaptation. To examine the



Global Trade Item Number

SKUGTIN
344906-1ML04055977214925

Questions

Reviews

No rating value

Active Filters