Skip to Content
Merck
All Photos(1)

Key Documents

205530

Sigma-Aldrich

CA-074

CA-074, also referenced under CAS 134448-10-5, modulates the biological activity of cathepsin B. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

Synonym(s):

CA-074, Cathepsin B Inhibitor III, [L-3- trans-(Propylcarbamoyl)oxirane-2-carbonyl]-L-isoleucyl-L-proline

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
CZK 6,150.00

CZK 6,150.00


Estimated to ship onApril 22, 2025Details



Select a Size

Change View
1 MG
CZK 6,150.00

About This Item

Empirical Formula (Hill Notation):
C18H29N3O6
Molecular Weight:
383.44
UNSPSC Code:
12352200
NACRES:
NA.77

CZK 6,150.00


Estimated to ship onApril 22, 2025Details


Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

colorless to white

solubility

DMSO: 10 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A potent, irreversible inhibitor of Cathepsin B in vivo and in vitro (IC50 = 2.24 nM for rat liver cathepsin B).
An epoxysuccinyl peptide that acts as a potent, irreversible, and specific inhibitor of cathepsin B (IC50 = 2.24 nM for rat liver cathepsin B). Reduces ischemia-induced neuronal death. Exhibits lower membrane permeability when compared to CA-074 Me (Cat. No. 205531).

Biochem/physiol Actions

Cell permeable: no
Primary Target
cathepsin B
Product does not compete with ATP.
Reversible: no
Target IC50: 2.24 nM for rat liver cathepsin B

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Chan, V.J., et al. 1999. Biochem. J. 340, 113.
Tsuchiya, K., et al. 1999. Exp. Neurol. 155, 187.
Yamamoto, A., et al. 1997. J. Biochem.121, 974.
Inubushi, T., et al. 1994. J. Biochem. 116, 282.
Murata, M., et al. 1991. FEBS Lett. 280, 307.
Towatari, T., et al. 1991. FEBS Lett. 280, 311.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anneliese S Ashhurst et al.
bioRxiv : the preprint server for biology (2021-01-06)
The emergence of SARS-CoV-2 in late 2019, and the subsequent COVID-19 pandemic, has led to substantial mortality, together with mass global disruption. There is an urgent need for novel antiviral drugs for therapeutic or prophylactic application. Cathepsin L is a
Ananda Baral et al.
International journal of molecular sciences, 22(22) (2021-11-28)
Leptin, a hormone that is predominantly produced by adipose tissue, is closely associated with various liver diseases. However, there is a lack of understanding as to whether leptin directly induces cytotoxic effects in hepatocytes as well as the mechanisms that

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service