06-886-I
Anti-NFκB p50 Antibody
from rabbit, purified by affinity chromatography
Synonym(s):
Nuclear factor NF-kappa-B p105 subunit, NFκB p50, DNA-binding factor KBF1, EBP-1, Nuclear factor of kappa light polypeptide gene enhancer in B-cells 1
Select a Size
CZK 10,700.00
Estimated to ship onApril 11, 2025Details
Select a Size
About This Item
CZK 10,700.00
Estimated to ship onApril 11, 2025Details
Recommended Products
biological source
rabbit
Quality Level
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
purified by
affinity chromatography
species reactivity
human
species reactivity (predicted by homology)
rhesus macaque (based on 100% sequence homology)
packaging
antibody small pack of 25 μg
technique(s)
immunoprecipitation (IP): suitable
western blot: suitable
NCBI accession no.
UniProt accession no.
shipped in
ambient
target post-translational modification
unmodified
Gene Information
human ... NFKB1(4790)
General description
Specificity
Immunogen
Application
Quality
Western Blotting Analysis: 0.2 µg/mL of this antibody detected NFkB p50 in 10 µg of A431 cell lysate.
Target description
Other Notes
Not finding the right product?
Try our Product Selector Tool.
recommended
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service