About This Item
Product Name
Acetyl-Histone H3 (Lys9) Peptide, Routinely evaluated in a Dot Blot Assay by spotting on nitrocellulose & detecting by using a Anti-acetyl Histone H3 (Lys9) (06-942).
Quality Level
manufacturer/tradename
Upstate®
technique(s)
western blot: suitable
NCBI accession no.
UniProt accession no.
shipped in
dry ice
Gene Information
human ... H3F3B(3021)
Analysis Note
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| technique(s) western blot: suitable | technique(s) immunocytochemistry: suitable, western blot: suitable | technique(s) immunocytochemistry: suitable, immunoprecipitation (IP): suitable, western blot: suitable | technique(s) activity assay: suitable (histone acetyltransferase (HAT)), western blot: suitable |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
| UniProt accession no. | UniProt accession no. | UniProt accession no. | UniProt accession no. |
| shipped in dry ice | shipped in wet ice | shipped in dry ice | shipped in dry ice |
| Gene Information human ... H3F3B(3021) | Gene Information human ... H3F3B(3021) | Gene Information human ... H3F3B(3021) | Gene Information - |
| manufacturer/tradename Upstate® | manufacturer/tradename Upstate® | manufacturer/tradename Upstate® | manufacturer/tradename Upstate® |
Still not finding the right product?
Explore all of our products under Acetyl-Histone H3 (Lys9) Peptide
Storage Class
11 - Combustible Solids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 12-358 | 04053252477591 |



