Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
100 μg | Estimated to ship onJune 08, 2026fromAreál Kühne+Nagel spol. s r.o. | CZK 8,150.00 |
About This Item
Quality Segment
form
liquid
manufacturer/tradename
Upstate®
technique(s)
activity assay: suitable (histone acetyltransferase (HAT)), western blot: suitable
NCBI accession no.
UniProt accession no.
shipped in
dry ice
Application
Physical form
Preparation Note
Analysis Note
Legal Information
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| technique(s) activity assay: suitable (histone acetyltransferase (HAT)), western blot: suitable | technique(s) activity assay: suitable (histone acetyltransferase (HAT)) | technique(s) activity assay: suitable (histone acetyltransferase (HAT)), western blot: suitable | technique(s) activity assay: suitable (histone acetyltransferase (HAT)) |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
| form liquid | form liquid | form - | form liquid |
| UniProt accession no. | UniProt accession no. | UniProt accession no. | UniProt accession no. |
| shipped in dry ice | shipped in dry ice | shipped in dry ice | shipped in dry ice |
| manufacturer/tradename Upstate® | manufacturer/tradename Upstate® | manufacturer/tradename Upstate® | manufacturer/tradename Upstate® |
Storage Class
12 - Non Combustible Liquids
wgk
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



