Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|---|---|---|
| 100 μg | Please contact Customer Service for Availability | CZK 7,930.00 |
About This Item
UNSPSC Code:
12352200
NACRES:
NA.41
eCl@ss:
32160405
Quality Segment
manufacturer/tradename
Upstate®
technique(s)
activity assay: suitable
NCBI accession no.
UniProt accession no.
shipped in
dry ice
Gene Information
human ... H3F3B(3021)
Physical form
Lyophilized powder
Preparation Note
2 years at -20°C
Analysis Note
Routinely evaluated in a Dot Blot Assay by spotting on nitrocellulose and detecting with polyclonal anti-Histone H3 (06-755).
Legal Information
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
1 of 1
This Item | |||
|---|---|---|---|
| technique(s) activity assay: suitable | technique(s) immunocytochemistry: suitable, western blot: suitable | technique(s) western blot: suitable | technique(s) immunoprecipitation (IP): suitable |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
| shipped in dry ice | shipped in wet ice | shipped in dry ice | shipped in wet ice |
| UniProt accession no. | UniProt accession no. | UniProt accession no. | UniProt accession no. |
| Gene Information human ... H3F3B(3021) | Gene Information human ... H3F3B(3021) | Gene Information human ... H3F3B(3021) | Gene Information human ... H3F3B(3021) |
| manufacturer/tradename Upstate® | manufacturer/tradename Upstate® | manufacturer/tradename Upstate® | manufacturer/tradename Upstate® |
Still not finding the right product?
Explore all of our products under Histone H3 Peptide
Storage Class
11 - Combustible Solids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 12-357 | 04053252362101 |



