Skip to Content
Merck
All Photos(1)

Key Documents

1.04022

Sigma-Aldrich

Fast green FCF (C.I. 42053)

for microscopy Certistain®

Synonym(s):

Fast green FCF (C.I. 42053), (C.I. 42053) for microbiology Certistain®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H34N2Na2O10S3
CAS Number:
Molecular Weight:
808.85
MDL number:
UNSPSC Code:
41116130
EC Index Number:
219-091-5

Quality Level

form

solid

potency

>2000 mg/kg LD50, oral (Rat)

composition

Dye, ≥85% spectrophotometric assay

pH

2.7 (20 °C, 10 g/L in H2O)

mp

290 °C (decomposition)

solubility

150 g/L

bulk density

350 kg/m3

εmax

1360-1610 at 622-626 nm in 50% ethanol at 0.003 g/L

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-30°C

InChI

1S/C37H36N2O10S3.2Na/c1-3-38(24-26-7-5-9-33(21-26)50(41,42)43)30-15-11-28(12-16-30)37(35-20-19-32(40)23-36(35)52(47,48)49)29-13-17-31(18-14-29)39(4-2)25-27-8-6-10-34(22-27)51(44,45)46;;/h5-23H,3-4,24-25H2,1-2H3,(H3-,40,41,42,43,44,45,46,47,48,49);;/q;2*+1/p-2

InChI key

XJBPDZVCYTYRIN-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

The dry dye Fast Green FCF (C.I. 42053) for microscopy Certistain®, is used for the preparation of a staining solution that can be used for modified connective tissue staining acc. to van Gieson in histological sections. Further it is applied in histology as counterstaining in e.g. trichrome staining according to Masson and additionally for histochemical staining, as well as in Cytology as counterstain instead of light green SF in Papanicolaou′s staining. As Certistain® dye it is chemically quality-certified according to highest standards and strict specifications. This product is registered as IVD and CE marked. For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.

Analysis Note

Dye content (spectrophotometrically): ≥ 85 %
Identity (UV/VIS-Spectrum): passes test
Absorption maximum λ max (ethanol 50 %): 622 - 626 nm
Spec. Absorptivity A 1%/1cm (λmax; 0.003 g/l; ethanol 50 %): 1360 - 1610
TLC-Test: passes test
Loss on drying (110 °C): ≤ 10 %
Suitability for microscopy: passes test

Legal Information

CERTISTAIN is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Muta. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service