Skip to Content
Merck

Skip To

07-807

Anti-phospho-Met (Tyr1003) Antibody

Upstate®, from rabbit

Synonym(s):

c-Met, cmet, HGFR, RCCP2, met proto-oncogene (hepatocyte growth factor receptor)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
100 μL

Estimated to ship onMay 29, 2026fromAreál Kühne+Nagel spol. s r.o.

CZK 10,800.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Clone:
polyclonal
Species reactivity:
human
Application:
WB
Citations:
2

CZK 10,800.00


Estimated to ship onMay 29, 2026Details


Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

phosphorylation (pTyr1003)

Gene Information

human ... MET(4233)

General description

155 kDa

Immunogen

Peptide corresponding to amino acid region encompassing the human, mouse, and rat phospho-Met (Tyr1003).

Application

Anti-phospho-Met (Tyr1003) Antibody is an antibody against phospho-Met (Tyr1003) for use in WB.

Biochem/physiol Actions

Predicted to cross-react with Mouse and Rat based on complete sequence homology.
Recognizes Met phosphorylated on Tyr1003.

Analysis Note

Routinely evaluated by immunoblot.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
07-81005-1049SAB4300599
species reactivity

human

species reactivity

mouse, human

species reactivity

human

species reactivity

rat, human, mouse

antibody form

affinity purified immunoglobulin

antibody form

affinity purified immunoglobulin

antibody form

purified antibody

antibody form

affinity isolated antibody

clone

polyclonal

clone

polyclonal

clone

4F8.2, monoclonal

clone

polyclonal

biological source

rabbit

biological source

rabbit

biological source

mouse

biological source

rabbit

UniProt accession no.

P08581

UniProt accession no.

P08581

UniProt accession no.

P08581

UniProt accession no.

P08581

shipped in

dry ice

shipped in

dry ice

shipped in

wet ice

shipped in

wet ice


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 2



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
07-80704053252735264

Questions

Reviews

No rating value

Active Filters