Skip to Content
Merck

Skip To

07-289

Anti-acetyl-Histone H2A (Lys9) Antibody

serum, Upstate®

Synonym(s):

H2AK9Ac, Histone H2A (acetyl K9)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
200 μL

Estimated to ship onMay 19, 2026fromAreál Kühne+Nagel spol. s r.o.

CZK 14,300.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Clone:
polyclonal
Species reactivity:
vertebrates, human
Application:
WB
Citations:
5

CZK 14,300.00


Estimated to ship onMay 19, 2026Details


Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

vertebrates, human

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

acetylation (Lys9)

General description

14kDa

Immunogen

Peptide (GKQGGAcKARAKAK-C) corresponding to amino acids 4-15 of human Histone H2A.

Application

Anti-acetyl-Histone H2A (Lys9) Antibody is a Rabbit Polyclonal Antibody for detection of acetyl-Histone H2A (Lys9) also known as H2AK9Ac, Histone H2A (acetyl K9) & has been validated in WB.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones

Biochem/physiol Actions

acetylated Histone 2A

Physical form

Antiserum
antiserum immuno-depleted with the non-acetylated immunogenic peptide, containing 0.05% sodium azide and 30% glycerol

Preparation Note

2 years at -20°C

Analysis Note

routinely evaluated by immunoblot on sodium butyrate treated acid extracted proteins from HeLa cells

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
07-14607-32806-942
clone

polyclonal

clone

polyclonal

clone

polyclonal

clone

polyclonal

antibody form

serum

antibody form

serum

antibody form

serum

antibody form

purified immunoglobulin

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

species reactivity

vertebrates, human

species reactivity

yeast, chicken, human, rat, Saccharomyces cerevisiae, mouse

species reactivity

human, Saccharomyces cerevisiae

species reactivity

mouse, human, rat

shipped in

dry ice

shipped in

dry ice

shipped in

wet ice

shipped in

dry ice

technique(s)

western blot: suitable

technique(s)

ChIP: suitable, western blot: suitable

technique(s)

ChIP: suitable (ChIP-seq), dot blot: suitable, immunoprecipitation (IP): suitable, western blot: suitable

technique(s)

ChIP: suitable (ChIP-seq), dot blot: suitable, flow cytometry: suitable, western blot: suitable


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 1



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content

Cancer is a complex disease manifestation. At its core, it remains a disease of abnormal cellular proliferation and inappropriate gene expression. In the early days, carcinogenesis was viewed simply as resulting from a collection of genetic mutations that altered the gene expression of key oncogenic genes or tumor suppressor genes leading to uncontrolled growth and disease (Virani, S et al 2012). Today, however, research is showing that carcinogenesis results from the successive accumulation of heritable genetic and epigenetic changes. Moreover, the success in how we predict, treat and overcome cancer will likely involve not only understanding the consequences of direct genetic changes that can cause cancer, but also how the epigenetic and environmental changes cause cancer (Johnson C et al 2015; Waldmann T et al 2013). Epigenetics is the study of heritable gene expression as it relates to changes in DNA structure that are not tied to changes in DNA sequence but, instead, are tied to how the nucleic acid material is read or processed via the myriad of protein-protein, protein-nucleic acid, and nucleic acid-nucleic acid interactions that ultimately manifest themselves into a specific expression phenotype (Ngai SC et al 2012, Johnson C et al 2015). This review will discuss some of the principal aspects of epigenetic research and how they relate to our current understanding of carcinogenesis. Because epigenetics affects phenotype and changes in epigenetics are thought to be key to environmental adaptability and thus may in fact be reversed or manipulated, understanding the integration of experimental and epidemiologic science surrounding cancer and its many manifestations should lead to more effective cancer prognostics as well as treatments (Virani S et al 2012).

Histone Modifications Poster


Tanja Waldmann et al.
Epigenetics & chromatin, 4, 11-11 (2011-07-22)
Covalent histone modifications are central to all DNA-dependent processes. Modifications of histones H3 and H4 are becoming well characterised, but knowledge of how H2A modifications regulate chromatin dynamics and gene expression is still very limited. To understand the function of
Michael S Kobor et al.
PLoS biology, 2(5), E131-E131 (2004-03-27)
The conserved histone variant H2A.Z functions in euchromatin to antagonize the spread of heterochromatin. The mechanism by which histone H2A is replaced by H2A.Z in the nucleosome is unknown. We identified a complex containing 13 different polypeptides associated with a
Bin Wang et al.
Nature communications, 14(1), 3371-3371 (2023-06-09)
In the Neurospora circadian system, the White Collar Complex (WCC) drives expression of the principal circadian negative arm component frequency (frq). FRQ interacts with FRH (FRQ-interacting RNA helicase) and CKI, forming a stable complex that represses its own expression by inhibiting



Global Trade Item Number

SKUGTIN
07-28904053252369056

Questions

Reviews

No rating value

Active Filters