Skip to Content
Merck
All Photos(1)

Key Documents

05-479

Sigma-Aldrich

Anti-Cytochrome C Antibody, clone C-7

ascites fluid, clone C-7, Upstate®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

mouse

antibody form

ascites fluid

antibody product type

primary antibodies

clone

C-7, monoclonal

species reactivity

human, horse

manufacturer/tradename

Upstate®

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... CYCS(54205)

Specificity

Cytochrome c

Immunogen

Horse cytochrome c conjugated to KLH

Application

Detect Cytochrome C using this Anti-Cytochrome C Antibody, clone C-7 validated for use in WB, IH.
Research Category
Apoptosis & Cancer

Metabolism
Research Sub Category
Apoptosis - Additional

Enzymes & Biochemistry

Quality

routinely evaluated by immunoblot on RIPA lysates from A431 cells

Target description

14kDa

Linkage

Replaces: 04-1043

Physical form

Ascites
mouse ascites containing 0.05% sodium azide

Storage and Stability

2 years at -20°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structural basis for the binding of an anti-cytochrome c antibody to its antigen: crystal structures of FabE8-cytochrome c complex to 1.8 A resolution and FabE8 to 2.26 A resolution.
Mylvaganam, S E, et al.
Journal of Molecular Biology, 281, 301-322 (1998)
J E Springer et al.
Nature medicine, 5(8), 943-946 (1999-07-30)
Traumatic spinal cord injury often results in complete loss of voluntary motor and sensory function below the site of injury. The long-term neurological deficits after spinal cord trauma may be due in part to widespread apoptosis of neurons and oligodendroglia
Rui Liu et al.
Molecular cancer therapeutics, 14(9), 2090-2102 (2015-07-05)
Tumor adaptive resistance to therapeutic radiation remains a barrier for further improvement of local cancer control. SIRT3, a member of the sirtuin family of NAD(+)-dependent protein deacetylases in mitochondria, promotes metabolic homeostasis through regulation of mitochondrial protein deacetylation and plays
So-Yeon Kim et al.
Oncotarget, 8(39), 64964-64973 (2016-08-05)
Small molecules to selectively induce cell death of undifferentiated human pluripotent stem cells (hPSCs) have been developed with the aim of lowering the risk of teratoma formation during hPSC-based cell therapy. In this context, we have reported that Quercetin (QC)
Indraneel Mittra et al.
Journal of biosciences, 40(1), 91-111 (2015-03-06)
Whether nucleic acids that circulate in blood have any patho-physiological functions in the host have not been explored.We report here that far from being inert molecules, circulating nucleic acids have significant biological activities of their own that are deleterious to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service