Skip to Content
Merck
All Photos(1)

Key Documents

MABT1363M

Sigma-Aldrich

Anti-GM130 Antibody, clone 4A3

clone 4A3, from mouse

Synonym(s):

Golgin subfamily A member 2, 130 kDa cis-Golgi matrix protein

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
CZK 3,890.00
200 μL
CZK 10,300.00

CZK 3,890.00


Estimated to ship onMay 30, 2025Details


Request a Bulk Order

Select a Size

Change View
25 μL
CZK 3,890.00
200 μL
CZK 10,300.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

CZK 3,890.00


Estimated to ship onMay 30, 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4A3, monoclonal

species reactivity

human, rat

packaging

antibody small pack of 25 μL

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable (paraffin)
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

General description

Golgin subfamily A member 2 (UniProt: Q62839; also known as 130 kDa cis-Golgi matrix protein, GM130) is encoded by the Golga2 gene (Gene ID: 64528) in rat. GM130 is a peripheral membrane component of the cis-Golgi stack that acts as a membrane skeleton to maintain the structure of the Golgi apparatus, and as a vesicle tether that facilitates vesicle fusion to the Golgi membrane. GM130 is a homotetramer with a flexible rod-like structure. It has a nuclear localization sequence (aa 26-49) that mediates interaction with importin-alpha and recruits it to the Golgi membrane and liberates TPX2. GM130 promotes the mitotic spindle pole assembly by activating TPX2 to nucleate microtubules around the Golgi and capture them to couple mitotic membranes to the spindle. Phosphorylation of GM130 at Ser37 by CDK1 at the onset of mitosis inhibits its interaction with p115/USO1, which prevents its tethering of COPI vesicles and thereby inhibits transport through the Golgi apparatus during mitosis.

Specificity

Clone 4A3 detected Golgin subfamily A member 2 (GM130) in human liver cancer and human intestine sections.

Immunogen

A full length recombinant human Golgin subfamily A member 2 (GM130).

Application

Anti-GM130, clone 4A3, Cat. No. MABT1363, is a mouse monoclonal antibody that detects Golgin subfamily A member 2. (GM130) and is tested for use in Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, and Western Blotting.
Immunohistochemistry Analysis: A 1:50 dilution of this antibody detected GM130 in human liver cancer tissue.

Western Blotting Analysis: A representative lot detected GM130 in Western Blotting applications (Diao, A., et. al. (2003) J Cell Biol. 160(2):201-12).

Immunocytochemistry Analysis: A representative lot detected GM130 in Immunocytochemistry applications (Seemann, J., et. al. (2002). Science. 295(5556):848-51; Diao, A., et. al. (2003) J Cell Biol. 160(2):201-12).

Immunohistochemistry Analysis: A representative lot detected GM130 in Immunohistochemistry applications (Seemann, J., et. al. (2002). Science. 295(5556):848-51).

Immunoprecipitation Analysis: A representative lot detected GM130 in Immunoprecipitation applications (Diao, A., et. al. (2003) J Cell Biol. 160(2):201-12; Diao, A., et. al. (2008) J Cell Biol. 283(11):6957-67).
Research Category
Cell Structure

Quality

Evaluated by Immunohistochemistry in human small intestine tissue.

Immunohistochemistry Analysis: A 1:50 dilution of this antibody detected GM130 in human small intestine tissue sections.

Target description

112.87 kDa calculated.

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service